missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sebacate, 97%

Catalog No. AAL1580522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1580522 100 g
AAL1580536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1580522 Supplier Thermo Scientific Chemicals Supplier No. L1580522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 106-79-6 | C12H22O4 | 230.304 g/mol

Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 106-79-6
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00008472
InChI Key ALOUNLDAKADEEB-UHFFFAOYSA-N
Synonym dimethyl sebacate, methyl sebacate, decanedioic acid, dimethyl ester, sebacic acid dimethyl ester, sebacic acid, dimethyl ester, dimethyl octane-1,8-dicarboxylate, 1,10-dimethyl decanedioate, decanedioic acid dimethyl ester, decanedioic acid, 1,10-dimethyl ester, dimethyl decane-1,10-dioate
PubChem CID 7829
IUPAC Name dimethyl decanedioate
SMILES COC(=O)CCCCCCCCC(=O)OC

Specifications

Melting Point 25°C to 28°C
Density 0.988
Boiling Point 287°C to 289°C
Flash Point 145°C (293°F)
Quantity 100 g
Beilstein 1785523
Solubility Information Insoluble in water.
Formula Weight 230.31
Percent Purity 97%
Chemical Name or Material Dimethyl sebacate
EINECSNumber 203-431-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only