missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (S)-(+)-2-Methylglutarate 97.0+%, TCI America™

Catalog No. M12305G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12305G Supplier TCI America Supplier No. M12305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10171-92-3
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00671564
InChI Key ZWKKRUNHAVNSFW-UHFFFAOYNA-N
Synonym (S)-(+)-2-Methylglutaric Acid Dimethyl Ester
PubChem CID 11052138
IUPAC Name 1,5-dimethyl 2-methylpentanedioate
SMILES COC(=O)CCC(C)C(=O)OC

Specifications

Color Colorless
Boiling Point 91°C
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (S)-(+)-2-Methylglutarate