missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Pimelimidate Dihydrochloride 98.0+%, TCI America™

Catalog No. P08925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08925G 5 g
P089225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08925G Supplier TCI America Supplier No. P08925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58537-94-3
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD00038942
InChI Key LRPQMNYCTSPGCX-UHFFFAOYSA-N
Synonym dimethyl pimelimidate dihydrochloride, pimelimidic acid dimethyl ester dihydrochloride, acmc-209tih, dimethyl pimelinediimidate dihydrochloride, dimethyl pimelinediimidate dihydro-chloride, dimethyl heptanebis imidate dihydrochloride, dimethyl pimelimidate dihydrochloride, powder, dimethyl pimelinediimidate paragraph signthornnieani, dimethyl pimelimidate dihydrochloride at
PubChem CID 11402688
IUPAC Name 1,7-dimethyl heptanediimidate
SMILES COC(=N)CCCCCC(=N)OC

Specifications

Color White
Quantity 5 g
Formula Weight 259.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Pimelimidate Dihydrochloride [Cross-linking Agent for Peptides Research]
TSCA No
Recommended Storage Refrigerator