missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Phthalate 99.0+%, TCI America™
Supplier: TCI America P0302500G
Specifications
| Dimethyl Phthalate | |
| Colorless | |
| C10H10O4 | |
| 500 g | |
| NIQCNGHVCWTJSM-UHFFFAOYSA-N | |
| 1,2-dimethyl benzene-1,2-dicarboxylate | |
| 8554 | |
| 194.19 | |
| Liquid |
| 131-11-3 | |
| 284°C | |
| MFCD00008425 | |
| dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm | |
| COC(=O)C1=CC=CC=C1C(=O)OC | |
| 194.19 | |
| CHEBI:4609 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 131-11-3 | |
| 194.19 | |
| NIQCNGHVCWTJSM-UHFFFAOYSA-N | |
| 8554 | |
| 1,2-dimethyl benzene-1,2-dicarboxylate |
| C10H10O4 | |
| MFCD00008425 | |
| dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm | |
| CHEBI:4609 | |
| COC(=O)C1=CC=CC=C1C(=O)OC |
Safety and Handling
EINECSNumber : (3)-1301
RTECSNumber : TI1575000
TSCA : Yes