missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate 99.0+%, TCI America™

Catalog No. p-7134545
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
P030225G 25 g
P0302500G 500 g
2 options

Catalog No. P030225G

Supplier: TCI America P030225G

Only null left

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
IUPAC Name 1,2-dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Color Colorless
Boiling Point 284°C
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Phthalate
EINECSNumber (3)-1301
RTECSNumber TI1575000
TSCA Yes