missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phenylphosphonite, 98%

CAS: 2946-61-4 | C8H11O2P | 170.15 g/mol

Supplier:  Thermo Scientific Chemicals A1400603

 View more versions of this product

Catalog No. AAA1400603


Only null left
Explore available promotions

Description

Description

Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Dimethyl phenylphosphonite
1.072
C8H11O2P
MFCD00008352
1073018
dimethyl phenylphosphonite, dimethoxyphenylphosphine, phenyldimethoxyphosphine, dimethylphenylphosphonite, phosphorus, dimethoxy phenyl, phosphonous acid, phenyl-, dimethyl ester, phenylphosphonous acid dimethyl ester, acmc-20al6n, php ome 2, dimethoxy phenyl phosphane
LMZLQYYLELWCCW-UHFFFAOYSA-N
dimethoxy(phenyl)phosphane
76277
98%
2946-61-4
>110°C (230°F)
1.529
1 g
Air sensitive
Miscible with alcohol. Immiscible with water.
COP(OC)C1=CC=CC=C1
170.15
170.15

Chemical Identifiers

2946-61-4
170.15
LMZLQYYLELWCCW-UHFFFAOYSA-N
76277
COP(OC)C1=CC=CC=C1
C8H11O2P
MFCD00008352
dimethyl phenylphosphonite, dimethoxyphenylphosphine, phenyldimethoxyphosphine, dimethylphenylphosphonite, phosphorus, dimethoxy phenyl, phosphonous acid, phenyl-, dimethyl ester, phenylphosphonous acid dimethyl ester, acmc-20al6n, php ome 2, dimethoxy phenyl phosphane
dimethoxy(phenyl)phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 220-960-6

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only