missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phenylphosphonite, 98%

Catalog No. p-7021604
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAA1400603 1 g
AAA1400606 5 g
AAA1400614 25 g
3 options

Catalog No. AAA1400603

Supplier: Thermo Scientific Chemicals A1400603

Only null left

CAS: 2946-61-4 | C8H11O2P | 170.15 g/mol

Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2946-61-4
Molecular Formula C8H11O2P
Molecular Weight (g/mol) 170.15
MDL Number MFCD00008352
InChI Key LMZLQYYLELWCCW-UHFFFAOYSA-N
Synonym dimethyl phenylphosphonite, dimethoxyphenylphosphine, phenyldimethoxyphosphine, dimethylphenylphosphonite, phosphorus, dimethoxy phenyl, phosphonous acid, phenyl-, dimethyl ester, phenylphosphonous acid dimethyl ester, acmc-20al6n, php ome 2, dimethoxy phenyl phosphane
PubChem CID 76277
IUPAC Name dimethoxy(phenyl)phosphane
SMILES COP(OC)C1=CC=CC=C1

Specifications

Density 1.072
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Beilstein 1073018
Sensitivity Air sensitive
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 170.15
Percent Purity 98%
Chemical Name or Material Dimethyl phenylphosphonite
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 220-960-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only