missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Nitroterephthalate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 75°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 239.18 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethyl Nitroterephthalate |
Safety and Handling
| TSCA | Yes |