missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl malonate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 159560025

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Catalog No. AC159560025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

108-59-8
132.12
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
dimethyl propanedioate
C5H8O4
BEPAFCGSDWSTEL-UHFFFAOYSA-N
7943
COC(=O)CC(=O)OC

Specifications

Dimethyl malonate
1.1560g/mL
90°C
96% min. (GC)
C5H8O4
CH3O2CCH2CO2CH3
02,572
15,6168
dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
BEPAFCGSDWSTEL-UHFFFAOYSA-N
dimethyl propanedioate
7943
97%
108-59-8
180.0°C to 181.0°C
Authentic
Glass bottle
1.4120 to 1.4140
2.5L
08,193; 13,34
1.156
Solubility in water: negligible. Other solubilities: soluble in alcohol and ether
COC(=O)CC(=O)OC
132.12
132.12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-597-8

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...