missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl malonate, 97%, Thermo Scientific™

Catalog No. p-4258976
Change view
Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159562500 250mL Glass bottle
AC159560010 1L Glass bottle
AC159560025 2.5L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC159562500 Supplier Thermo Scientific Chemicals Supplier No. 159562500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 108-59-8
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
InChI Key BEPAFCGSDWSTEL-UHFFFAOYSA-N
Synonym dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
PubChem CID 7943
IUPAC Name dimethyl propanedioate
SMILES COC(=O)CC(=O)OC

Specifications

Density 1.1560g/mL
Boiling Point 180.0°C to 181.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140
Linear Formula CH3O2CCH2CO2CH3
Quantity 250mL
Beilstein 02,572
Fieser 08,193; 13,34
Merck Index 15,6168
Specific Gravity 1.156
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and ether
Formula Weight 132.12
Percent Purity 97%
Chemical Name or Material Dimethyl malonate
Show More Show Less
EINECSNumber 203-597-8

RUO – Research Use Only