missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl malonate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 159560010
| Quantity | 1L |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 108-59-8 | |
| 132.12 | |
| dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
| dimethyl propanedioate |
| C5H8O4 | |
| BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
| 7943 | |
| COC(=O)CC(=O)OC |
Specifications
| Dimethyl malonate | |
| 1.1560g/mL | |
| 90°C | |
| 96% min. (GC) | |
| C5H8O4 | |
| CH3O2CCH2CO2CH3 | |
| 02,572 | |
| 15,6168 | |
| dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester | |
| BEPAFCGSDWSTEL-UHFFFAOYSA-N | |
| dimethyl propanedioate | |
| 7943 | |
| 97% |
| 108-59-8 | |
| 180.0°C to 181.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4120 to 1.4140 | |
| 1L | |
| 08,193; 13,34 | |
| 1.156 | |
| Solubility in water: negligible. Other solubilities: soluble in alcohol and ether | |
| COC(=O)CC(=O)OC | |
| 132.12 | |
| 132.12 |
Safety and Handling
EINECSNumber : 203-597-8
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.