missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl L-Aspartate Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America A150625G

Quantity 25 g

 View more versions of this product

Catalog No. A150625G


Only null left

Description

Description

Specifications

Chemical Identifiers

32213-95-9
197.615
PNLXWGDXZOYUKB-WCCKRBBISA-N
2734892
COC(=O)CC(C(=O)OC)N.Cl
C6H12ClNO4
MFCD00038878
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
dimethyl (2S)-2-aminobutanedioate;hydrochloride

Specifications

116°C
32213-95-9
C6H12ClNO4
h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
COC(=O)CC(C(=O)OC)N.Cl
25 g
2734892
≥98.0% (N,T)
Dimethyl L-Aspartate Hydrochloride
White
MFCD00038878
PNLXWGDXZOYUKB-WCCKRBBISA-N
dimethyl (2S)-2-aminobutanedioate;hydrochloride
197.615
197.62
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions