missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Itaconate (stabilized with HQ) 98.0+%, TCI America™
$49.36 - $208.85
Chemical Identifiers
| CAS | 617-52-7 |
|---|---|
| Molecular Formula | C7H10O4 |
| Molecular Weight (g/mol) | 158.15 |
| MDL Number | MFCD00008446 |
| InChI Key | ZWWQRMFIZFPUAA-UHFFFAOYSA-N |
| Synonym | dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester |
| PubChem CID | 69240 |
| IUPAC Name | 1,4-dimethyl 2-methylidenebutanedioate |
| SMILES | COC(=O)CC(=C)C(=O)OC |
Chemical Identifiers
| C7H10O4 | |
| MFCD00008446 | |
| dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester | |
| 1,4-dimethyl 2-methylidenebutanedioate |
Specifications
| 617-52-7 | |
| 109°C | |
| MFCD00008446 | |
| ZWWQRMFIZFPUAA-UHFFFAOYSA-N | |
| 1,4-dimethyl 2-methylidenebutanedioate | |
| 69240 | |
| ≥98.0% (GC) | |
| Dimethyl Itaconate (stabilized with HQ) |
| White-Yellow | |
| C7H10O4 | |
| dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester | |
| COC(=O)CC(=C)C(=O)OC | |
| 158.15 | |
| 158.15 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-1130&(2)-1142
TSCA : Yes