missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl itaconate, 97%

Catalog No. p-7045974
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1265222 100 g
AAL1265236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1265222 Supplier Thermo Scientific Chemicals Supplier No. L1265222
Only null left

CAS: 617-52-7 | C7H10O4 | 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 617-52-7
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00008446
InChI Key ZWWQRMFIZFPUAA-UHFFFAOYSA-N
Synonym dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester
PubChem CID 69240
IUPAC Name dimethyl 2-methylidenebutanedioate
SMILES COC(=O)CC(=C)C(=O)OC

Specifications

Melting Point 33°C to 39°C
Density 1.124
Boiling Point 207°C to 208°C
Flash Point 100°C (212°F)
Quantity 100 g
Beilstein 386674
Formula Weight 158.16
Percent Purity 97%
Chemical Name or Material Dimethyl itaconate
EINECSNumber 210-519-6
TSCA Yes

RUO – Research Use Only