missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isophthalate, 98%

Catalog No. p-7027206
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2377922 100 g
AAB2377936 500 g
AAB237790E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2377922 Supplier Thermo Scientific Chemicals Supplier No. B2377922
Only null left

CAS: 1459-93-4 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1459-93-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008433
InChI Key VNGOYPQMJFJDLV-UHFFFAOYSA-N
Synonym dimethyl isophthalate, dimethyl m-phthalate, isophthalic acid dimethyl ester, methyl isophthalate, 1,3-benzenedicarboxylic acid, dimethyl ester, morflex 1129, dimethyl 1,3-benzenedicarboxylate, dimethylisophthalate, methyl 3-carbomethoxy benzoate, isophthalic acid, dimethyl ester
PubChem CID 15088
IUPAC Name dimethyl benzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC=C1)C(=O)OC

Specifications

Melting Point 67°C to 70°C
Density 1.194
Boiling Point 282°C
Flash Point 138°C (280°F)
Odor Mild
Quantity 100 g
Beilstein 1912251
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material Dimethyl isophthalate
Hazard Category H320
Precautionary Statement P264b-P280-P305+P351+P338-P501c
EINECSNumber 215-951-9
RTECSNumber NT2540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only