missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Iminodiacetate Hydrochloride 98.0+%, TCI America™

Catalog No. D53855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53855G 5 g
D538525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53855G Supplier TCI America Supplier No. D53855G

Chemical Identifiers

CAS 39987-25-2
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.62
MDL Number MFCD00216724
InChI Key IIWYYIACSUPJCN-UHFFFAOYSA-N
Synonym dimethyl iminodiacetate hydrochloride, dimethyl 2,2'-azanediyldiacetate hydrochloride, methyl 2-2-methoxy-2-oxoethyl amino acetate hydrochloride, dimethyl iminodiacetic acid hydrochloride, iminodiacetic acid dimethyl ester hydrochloride, glycine, n-2-methoxy-2-oxoethyl-, methyl ester, hydrochloride, methyl 2-methoxycarbonyl methyl amino acetate, chloride, zlchem 797, acmc-1aeyr, iminodiacetic dimethyl ester hydrochloride
PubChem CID 2727727
IUPAC Name hydrogen methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate chloride
SMILES [H+].[Cl-].COC(=O)CNCC(=O)OC

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 197.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Iminodiacetate Hydrochloride