accessibility menu, dialog, popup

UserName

HClH3COOHNOOCH3

CAS RN 39987-25-2

IUPAC Name: hydrogen methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate chloride
Synonyms: hydrogen methyl 2-[(2-methoxy-2-oxoethyl)amino]acetate chloride
Molecular Weight (g/mol): 197.62
Molecular Formula: C6H12ClNO4
InChi Key: IIWYYIACSUPJCN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)CNCC(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results