missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Glutarate 98.0+%, TCI America™

Catalog No. G018525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G018525G 25 g
G0185100G 100 g
G0185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G018525G Supplier TCI America Supplier No. G018525G

Chemical Identifiers

CAS 1119-40-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00008468
InChI Key XTDYIOOONNVFMA-UHFFFAOYSA-N
Synonym dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
PubChem CID 14242
IUPAC Name 1,5-dimethyl pentanedioate
SMILES COC(=O)CCCC(=O)OC

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Glutarate
EINECSNumber (2)-0857&(2)-0925
TSCA Yes