missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Glutarate 98.0+%, TCI America™

Supplier:  TCI America G0185100G

Quantity 100 g

 View more versions of this product

Catalog No. G0185100G


Only null left

Description

Description

Specifications

Chemical Identifiers

1119-40-0
160.17
XTDYIOOONNVFMA-UHFFFAOYSA-N
14242
COC(=O)CCCC(=O)OC
C7H12O4
MFCD00008468
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
1,5-dimethyl pentanedioate

Specifications

Dimethyl Glutarate
Colorless
C7H12O4
MFCD00008468
XTDYIOOONNVFMA-UHFFFAOYSA-N
1,5-dimethyl pentanedioate
14242
≥98.0% (GC)
1119-40-0
214°C
100 g
dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
COC(=O)CCCC(=O)OC
160.17
160.17
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0857&(2)-0925

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions