missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl fluoromalonate, 97%

Catalog No. AAL1675106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1675106 5 g
AAL1675103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1675106 Supplier Thermo Scientific Chemicals Supplier No. L1675106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344-14-9 | C5H7FO4 | 150.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 344-14-9
Molecular Formula C5H7FO4
Molecular Weight (g/mol) 150.105
MDL Number MFCD00054680
InChI Key ZVXHZSXYHFBIEW-UHFFFAOYSA-N
PubChem CID 2737111
IUPAC Name dimethyl 2-fluoropropanedioate
SMILES COC(=O)C(C(=O)OC)F

Specifications

Boiling Point 111°C to 112°C (45 mmHg)
Refractive Index 1.4032
Quantity 5 g
UN Number UN3261
Beilstein 1768414
Synonym dimethyl 2-fluoromalonate, dimethyl fluoromalonate, dimethylfluoromalonate, dimethyl2-fluoromalonate, 2-fluoro-malonic acid dimethyl ester, 1,3-dimethyl 2-fluoropropanedioate, propanedioic acid, fluoro-, dimethyl ester, pubchem2148, dimethyl fluoropropanedioate, acmc-20a3w7
Formula Weight 150.11
Percent Purity 97%
Chemical Name or Material Dimethyl fluoromalonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only