accessibility menu, dialog, popup

UserName

CAS RN 344-14-9

IUPAC Name: 1,3-dimethyl 2-fluoropropanedioate
Synonyms: 1,3-dimethyl 2-fluoropropanedioate
Molecular Weight (g/mol): 150.11
Molecular Formula: C5H7FO4
InChi Key: ZVXHZSXYHFBIEW-UHFFFAOYSA-N
SMILES: COC(=O)C(F)C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results