missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Dodecanedioate 98.0+%, TCI America™
$96.94 - $557.35
Chemical Identifiers
| CAS | 1731-79-9 |
|---|---|
| Molecular Formula | C14H26O4 |
| Molecular Weight (g/mol) | 258.358 |
| MDL Number | MFCD00043632 |
| InChI Key | IZMOTZDBVPMOFE-UHFFFAOYSA-N |
| Synonym | dimethyl 1,10-decanedicarboxylate, dodecanedioic acid dimethyl ester, dodecanedioic acid, dimethyl ester, dimethyl 1,12-dodecanedioate, 1,12-dimethyl dodecanedioate, dodecanedioic acid dimethyl, dimethyl dodecane-1,12-dioate, dimethyldodecanedioate, dodecanedioic acid, 1,12-dimethyl ester, acmc-209e5w |
| PubChem CID | 74415 |
| IUPAC Name | dimethyl dodecanedioate |
| SMILES | COC(=O)CCCCCCCCCCC(=O)OC |
Chemical Identifiers
| 1731-79-9 | |
| 258.358 | |
| IZMOTZDBVPMOFE-UHFFFAOYSA-N | |
| 74415 | |
| COC(=O)CCCCCCCCCCC(=O)OC |
| C14H26O4 | |
| MFCD00043632 | |
| dimethyl 1,10-decanedicarboxylate, dodecanedioic acid dimethyl ester, dodecanedioic acid, dimethyl ester, dimethyl 1,12-dodecanedioate, 1,12-dimethyl dodecanedioate, dodecanedioic acid dimethyl, dimethyl dodecane-1,12-dioate, dimethyldodecanedioate, dodecanedioic acid, 1,12-dimethyl ester, acmc-209e5w | |
| dimethyl dodecanedioate |
Specifications
| 1731-79-9 | |
| C14H26O4 | |
| 25 g | |
| IZMOTZDBVPMOFE-UHFFFAOYSA-N | |
| dimethyl dodecanedioate | |
| 74415 | |
| ≥98.0% (GC) | |
| Dimethyl Dodecanedioate |
| White-Yellow | |
| MFCD00043632 | |
| dimethyl 1,10-decanedicarboxylate, dodecanedioic acid dimethyl ester, dodecanedioic acid, dimethyl ester, dimethyl 1,12-dodecanedioate, 1,12-dimethyl dodecanedioate, dodecanedioic acid dimethyl, dimethyl dodecane-1,12-dioate, dimethyldodecanedioate, dodecanedioic acid, 1,12-dimethyl ester, acmc-209e5w | |
| COC(=O)CCCCCCCCCCC(=O)OC | |
| 258.358 | |
| 258.36 | |
| Crystals |
Safety and Handling
EINECSNumber : (2)-0879
TSCA : Yes