missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Dodecanedioate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 258.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | Dimethyl Dodecanedioate |
Safety and Handling
| EINECSNumber | (2)-0879 |
| TSCA | Yes |