missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Decahydro-2,6-naphthalenedicarboxylate (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7127890
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37455G 5 g
D374525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37455G Supplier TCI America Supplier No. D37455G

Chemical Identifiers

CAS 3068-02-8
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.33
InChI Key XYHMJVZAEWACCS-UHFFFAOYNA-N
Synonym Decahydro-2,6-naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 11357248
IUPAC Name dimethyl 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene-2,6-dicarboxylate
SMILES COC(=O)C1CCC2CC(CCC2C1)C(=O)OC

Specifications

Color White
Boiling Point 140°C
Quantity 5 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Decahydro-2,6-naphthalenedicarboxylate (mixture of isomers)