missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl D-(-)-Tartrate 98.0+%, TCI America™

Catalog No. p-7136893
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16595G 5 g
T165925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16595G Supplier TCI America Supplier No. T16595G
Only null left

Chemical Identifiers

CAS 13171-64-7
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00008445
InChI Key PVRATXCXJDHJJN-IMJSIDKUSA-N
Synonym dimethyl d-tartrate, --dimethyl d-tartrate, dimethyl d---tartrate, dimethyl 2s,3s-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-, dimethyl ester, 2s,3s, d---tartaric acid dimethyl ester, dl-dimethyl tartrate, dl-dimethyl tartarate, d--dimethyl d-tartrate, meso-tartaric acid, dimethyl ester
PubChem CID 21142
IUPAC Name dimethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

Melting Point 60°C
Color White
Boiling Point 118°C
Quantity 5 g
Formula Weight 178.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl D-(-)-Tartrate