accessibility menu, dialog, popup

UserName

CAS RN 13171-64-7

IUPAC Name: 1,4-dimethyl (2S,3S)-2,3-dihydroxybutanedioate
Synonyms: 1,4-dimethyl (2S,3S)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: PVRATXCXJDHJJN-IMJSIDKUSA-N
SMILES: COC(=O)[C@@H](O)[C@H](O)C(=O)OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)