missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl D-tartrate, 99%

Catalog No. AAL1147206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1147206 5 g
AAL1147214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1147206 Supplier Thermo Scientific Chemicals Supplier No. L1147206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13171-64-7 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13171-64-7
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00008445
InChI Key PVRATXCXJDHJJN-IMJSIDKUSA-N
Synonym dimethyl d-tartrate, --dimethyl d-tartrate, dimethyl d---tartrate, dimethyl 2s,3s-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-, dimethyl ester, 2s,3s, d---tartaric acid dimethyl ester, dl-dimethyl tartrate, dl-dimethyl tartarate, d--dimethyl d-tartrate, meso-tartaric acid, dimethyl ester
PubChem CID 21142
IUPAC Name dimethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES COC(=O)C(C(C(=O)OC)O)O

Specifications

Melting Point 48°C to 56°C
Density 1.3
Boiling Point 158°C (11 mmHg)
Flash Point >170°C (338°F)
Quantity 5 g
Beilstein 1726254
Optical Rotation −21° (c=1 in Water)
Formula Weight 178.14
Percent Purity 99%
Chemical Name or Material Dimethyl D-tartrate
EINECSNumber 236-118-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only