missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl D-(+)-Malate 98.0+%, TCI America™
$300.11 - $999.13
Chemical Identifiers
| CAS | 70681-41-3 |
|---|---|
| Molecular Formula | C6H10O5 |
| Molecular Weight (g/mol) | 162.14 |
| MDL Number | MFCD00066216 |
| InChI Key | YSEKNCXYRGKTBJ-UHFFFAOYNA-N |
| Synonym | D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester |
| PubChem CID | 11062697 |
| IUPAC Name | 1,4-dimethyl 2-hydroxybutanedioate |
| SMILES | COC(=O)CC(O)C(=O)OC |
Chemical Identifiers
| 70681-41-3 | |
| 162.14 | |
| YSEKNCXYRGKTBJ-UHFFFAOYNA-N | |
| 11062697 | |
| COC(=O)CC(O)C(=O)OC |
| C6H10O5 | |
| MFCD00066216 | |
| D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester | |
| 1,4-dimethyl 2-hydroxybutanedioate |
Specifications
| 70681-41-3 | |
| C6H10O5 | |
| 5 g | |
| YSEKNCXYRGKTBJ-UHFFFAOYNA-N | |
| 1,4-dimethyl 2-hydroxybutanedioate | |
| 11062697 | |
| ≥98.0% (GC) | |
| Dimethyl D-(+)-Malate |
| Colorless | |
| MFCD00066216 | |
| D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester | |
| COC(=O)CC(O)C(=O)OC | |
| 162.14 | |
| 162.14 | |
| Liquid |
Safety and Handling
TSCA : No