missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Chloroacetal 98.0+%, TCI America™
$51.98 - $281.08
Chemical Identifiers
| CAS | 97-97-2 |
|---|---|
| Molecular Formula | C4H9ClO2 |
| Molecular Weight (g/mol) | 124.56 |
| MDL Number | MFCD00000948 |
| InChI Key | CRZJPEIBPQWDGJ-UHFFFAOYSA-N |
| Synonym | chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane |
| PubChem CID | 60986 |
| IUPAC Name | 2-chloro-1,1-dimethoxyethane |
| SMILES | COC(CCl)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C008525ML
|
TCI America
C008525ML |
25 mL |
Each for $51.98
|
|
|||||
|
C0085500ML
|
TCI America
C0085500ML |
500 mL |
Each for $281.08
|
|
|||||
Chemical Identifiers
| 97-97-2 | |
| 124.56 | |
| CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
| 60986 | |
| COC(CCl)OC |
| C4H9ClO2 | |
| MFCD00000948 | |
| chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
| 2-chloro-1,1-dimethoxyethane |
Specifications
| 97-97-2 | |
| 130°C | |
| MFCD00000948 | |
| 1993 | |
| CRZJPEIBPQWDGJ-UHFFFAOYSA-N | |
| 2-chloro-1,1-dimethoxyethane | |
| 60986 | |
| ≥98.0% (GC) | |
| Dimethyl Chloroacetal |
| Yellow | |
| C4H9ClO2 | |
| 25 mL | |
| chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane | |
| COC(CCl)OC | |
| 124.56 | |
| 124.56 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3589
TSCA : Yes