missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Carbonate 98.0+%, TCI America™

Catalog No. p-7124198
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C005325ML 25 mL
C0053100ML 100 mL
C0053500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C005325ML Supplier TCI America Supplier No. C005325ML
Only null left

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point 4°C
Color Colorless
Boiling Point 90°C
Quantity 25 mL
UN Number 1161
Formula Weight 90.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Carbonate
EINECSNumber (2)-2853
RTECSNumber FG0450000
TSCA Yes