missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Carbonate 98.0+%, TCI America™

Catalog No. c0053100ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
C0053100ML 100 mL
C005325ML 25 mL
C0053500ML 500 mL
3 options

Catalog No. C0053100ML

Supplier: TCI America C0053100ML

Only null left

Chemical Identifiers

CAS 616-38-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00008420
InChI Key IEJIGPNLZYLLBP-UHFFFAOYSA-N
Synonym methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192
PubChem CID 12021
ChEBI CHEBI:36596
IUPAC Name dimethyl carbonate
SMILES COC(=O)OC

Specifications

Melting Point 4°C
Color Colorless
Boiling Point 90°C
Quantity 100 mL
UN Number 1161
Formula Weight 90.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Carbonate
EINECSNumber (2)-2853
RTECSNumber FG0450000
TSCA Yes