missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Carbonate, 99%, Spectrum™ Chemical

Catalog No. 18610214
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610214 500 g Amber Glass Bottle
18610210 100 g Amber Glass Bottle
18610213 200 L Drum
18610211 2.5 kg Glass Bottle
18610212 20 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610214 Supplier Spectrum Chemical Mfg Cor Supplier No. D2285500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-38-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.99

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.