missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Carbonate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D2285200LTBL

 View more versions of this product

Catalog No. 18610213

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

616-38-6
Drum
0.99
1
200 L

Chemical Identifiers

616-38-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.