missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Bromoterephthalate 98.0+%, TCI America™

Catalog No. D50035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50035G 5 g
D500325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50035G Supplier TCI America Supplier No. D50035G
Only null left

Chemical Identifiers

CAS 18643-86-2
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00060638
InChI Key VUMPFOPENBVFOF-UHFFFAOYSA-N
Synonym Bromoterephthalic Acid Dimethyl Ester
PubChem CID 87741
IUPAC Name dimethyl 2-bromobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)Br

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 273.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl Bromoterephthalate