missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Azobenzene-4,4'-dicarboxylate 95.0+%, TCI America™

Catalog No. A15815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15815G 5 g
A15811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15815G Supplier TCI America Supplier No. A15815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5320-91-2
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.298
MDL Number MFCD00496622
InChI Key PSNNNVCIKCUWKM-UHFFFAOYSA-N
PubChem CID 585660
IUPAC Name methyl 4-[(4-methoxycarbonylphenyl)diazenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 240°C
Quantity 5 g
Synonym Azobenzene-4,4′-dicarboxylic Acid Dimethyl Ester, 4,4′-Azodibenzoic Acid Dimethyl Ester, Dimethyl 4,4′-Azodibenzoate
Formula Weight 298.30
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Azobenzene-4,4′-dicarboxylate