accessibility menu, dialog, popup

UserName

CAS RN 5320-91-2

IUPAC Name: methyl 4-{2-[4-(methoxycarbonyl)phenyl]diazen-1-yl}benzoate
Synonyms: methyl 4-{2-[4-(methoxycarbonyl)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol): 298.3
Molecular Formula: C16H14N2O4
InChi Key: PSNNNVCIKCUWKM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results