Learn More
Dimethyl aminoterephthalate, 99%
CAS: 5372-81-6 | C10H11NO4 | 209.201 g/mol
Supplier: Thermo Scientific Chemicals A105670B
Description
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Dimethyl aminoterephthalate | |
| 130°C to 134°C | |
| MFCD00008427 | |
| 2808505 | |
| Insoluble in water. | |
| COC(=O)C1=CC(=C(C=C1)C(=O)OC)N | |
| 209.201 | |
| 209.2 |
| 5372-81-6 | |
| C10H11NO4 | |
| 1000 g | |
| dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester | |
| DSSKDXUDARIMTR-UHFFFAOYSA-N | |
| dimethyl 2-aminobenzene-1,4-dicarboxylate | |
| 79336 | |
| 99% |
Chemical Identifiers
| 5372-81-6 | |
| 209.201 | |
| DSSKDXUDARIMTR-UHFFFAOYSA-N | |
| 79336 | |
| COC(=O)C1=CC(=C(C=C1)C(=O)OC)N |
| C10H11NO4 | |
| MFCD00008427 | |
| dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester | |
| dimethyl 2-aminobenzene-1,4-dicarboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-364-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only