missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl aminoterephthalate, 99%

Catalog No. AAA1056718
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1056718 50 g
AAA1056730 250 g
AAA105670B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1056718 Supplier Thermo Scientific Chemicals Supplier No. A1056718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5372-81-6 | C10H11NO4 | 209.201 g/mol

Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5372-81-6
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00008427
InChI Key DSSKDXUDARIMTR-UHFFFAOYSA-N
Synonym dimethyl 2-aminoterephthalate, dimethyl aminoterephthalate, dimethyl 3-aminoterephthalate, 1,4-benzenedicarboxylic acid, 2-amino-, dimethyl ester, aminoterephthalic acid dimethyl ester, 1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate, unii-91sf4e6i9w, 2-aminoterephthalic acid, dimethyl ester, 3-amino-4-methoxycarbonylbenzoic acid, methyl ester, terephthalic acid, amino-, dimethyl ester
PubChem CID 79336
IUPAC Name dimethyl 2-aminobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)C(=O)OC)N

Specifications

Melting Point 130°C to 134°C
Quantity 50 g
Beilstein 2808505
Solubility Information Insoluble in water.
Formula Weight 209.2
Percent Purity 99%
Chemical Name or Material Dimethyl aminoterephthalate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-364-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only