missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Adipimidate Dihydrochloride 98.0+%, TCI America™

Supplier:  TCI America A08065G

 View more versions of this product

Catalog No. A08065G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethyl Adipimidate Dihydrochloride [Cross-linking Agent for Protein Research]
215°C
C8H18Cl2N2O2
5 g
IEUUDEWWMRQUDS-UHFFFAOYSA-N
dimethyl hexanediimidate;dihydrochloride
12876176
≥98.0% (T)
14620-72-5
White
MFCD00012573
dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
COC(=N)CCCCC(=N)OC.Cl.Cl
245.144
245.14
Crystalline Powder

Chemical Identifiers

14620-72-5
245.144
IEUUDEWWMRQUDS-UHFFFAOYSA-N
12876176
COC(=N)CCCCC(=N)OC.Cl.Cl
C8H18Cl2N2O2
MFCD00012573
dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
dimethyl hexanediimidate;dihydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions