missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Adipimidate Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7117065
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08065G 5 g
A080625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08065G Supplier TCI America Supplier No. A08065G
Only null left

Chemical Identifiers

CAS 14620-72-5
Molecular Formula C8H18Cl2N2O2
Molecular Weight (g/mol) 245.144
MDL Number MFCD00012573
InChI Key IEUUDEWWMRQUDS-UHFFFAOYSA-N
Synonym dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
PubChem CID 12876176
IUPAC Name dimethyl hexanediimidate;dihydrochloride
SMILES COC(=N)CCCCC(=N)OC.Cl.Cl

Specifications

Melting Point 215°C
Color White
Quantity 5 g
Formula Weight 245.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Adipimidate Dihydrochloride [Cross-linking Agent for Protein Research]
TSCA No
Recommended Storage Refrigerator