missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl acetylenedicarboxylate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Specifications
| Melting Point | -18.0°C |
| Color | Colorless to Yellow |
| Density | 1.1600g/mL |
| Boiling Point | 95.0°C to 98.0°C (19.0 mmHg) |
| Flash Point | 86°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4460 to 1.4480 |
| Linear Formula | CH3O2CC≡CCO2CH3 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
| EINECSNumber | 212-098-4 |
RUO – Research Use Only