missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl acetylenedicarboxylate, 98%
Description
It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.
Solubility
It is insoluble in water but soluble in ethanol, ethyl ether, CCl4, miscible with organic solvents.
Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents, strong acids, strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
It acts as an inhibitor of oxidative phosphorylation. It is also used in Pharmaceutical and electronic industry. It is used in organic synthesis. It also acts as an intermediate in chemical research. It is also used in Diels-Alder Reaction.
Solubility
It is insoluble in water but soluble in ethanol, ethyl ether, CCl4, miscible with organic solvents.
Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents, strong acids, strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | -18°C |
| Density | 1.156 |
| Boiling Point | 95°C to 98°C (19 mmHg) |
| Flash Point | 86°C (186°F) |
| Refractive Index | 1.448 |
| Linear Formula | CH3O2CC≡CCO2CH3 |
| Quantity | 100 g |
| UN Number | UN1760 |
| Beilstein | 607063 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312+H332-H314 |
| Hazard Statement | GHS H Statement H314-H318-H302-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S. |
| EINECSNumber | 212-098-4 |
| RTECSNumber | ES0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only