missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetone-1,3-dicarboxylate, 97%

Catalog No. AAA1496914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1496914 25 g
AAA1496922 100 g
AAA1496936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1496914 Supplier Thermo Scientific Chemicals Supplier No. A1496914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Melting Point 16°C to 17°C
Density 1.185
Boiling Point 237°C to 241°C
Flash Point >110°C (230°F)
Odor Ester-like
Refractive Index 1.444
Quantity 25 g
Beilstein 1366042
IUPAC Name 1,5-dimethyl 3-oxopentanedioate
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Dimethyl acetone-1,3-dicarboxylate
Show More Show Less
missing translation for 'healthHazard1' H317
missing translation for 'healthHazard2' GHS H Statement
H317
May cause an allergic skin reaction.
missing translation for 'healthHazard3' P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 217-385-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only