missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate 95.0+%, TCI America™

Catalog No. D44141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44141G 1 g
D44145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44141G Supplier TCI America Supplier No. D44141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1829-60-3
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.185
InChI Key FQVSHUVBNGSEJO-UHFFFAOYSA-N
Synonym 7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylic Acid Dimethyl Ester, Dimethyl 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylate, 3,6-Epoxycyclohexa-1,4-diene-1,2-dicarboxylic Acid Dimethyl Ester
PubChem CID 12234536
IUPAC Name dimethyl 7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate
SMILES COC(=O)C1=C(C2C=CC1O2)C(=O)OC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 210.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 7-Oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate
TSCA No
Recommended Storage Refrigerator