missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-norbornene-2,3-dicarboxylate, 94%

Catalog No. AAL0866409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0866409 10 g
AAL0866418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0866409 Supplier Thermo Scientific Chemicals Supplier No. L0866409

CAS: 5826-73-3 | C11H14O4 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5826-73-3
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00154455
InChI Key VGQLNJWOULYVFV-UHFFFAOYNA-N
Synonym dimethyl 5-norbornene-2,3-dicarboxylate, dimethyl bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, dimethyl carbate, dimalone, nisy, compound 3,916, methyl 3-methoxycarbonyl bicyclo 2.2.1 hept-5-ene-2-carboxylate, acmc-1ah3y, norborn-5-ene-2endo,3endo-dicarboxylic acid dimethyl ester, 5-norbornene-2, dimethyl ester
PubChem CID 38295
IUPAC Name dimethyl bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES COC(=O)C1C2CC(C=C2)C1C(=O)OC

Specifications

Boiling Point 150°C to 152°C (20 mmHg)
Quantity 10 g
Beilstein 1248588
Merck Index 14,3240
Formula Weight 210.23
Percent Purity 94%
Chemical Name or Material Dimethyl 5-norbornene-2,3-dicarboxylate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
RTECSNumber RB8925000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only