missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-Hydroxyisophthalate 98.0+%, TCI America™

Catalog No. p-7130858
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H100725G 25 g
H1007250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H100725G Supplier TCI America Supplier No. H100725G

Chemical Identifiers

CAS 13036-02-7
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.19
MDL Number MFCD00134367
InChI Key DOSDTCPDBPRFHQ-UHFFFAOYSA-N
PubChem CID 83065
IUPAC Name 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(O)=C1)C(=O)OC

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Synonym dimethyl 5-hydroxyisophthalate, 5-hydroxyisophthalic acid dimethyl ester, 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester, dimethyl5-hydroxyisophthalate, timtec-bb sbb000491, 5-hydroxy-dimethylisophthalate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester, dimethyl-5-hydroxy-benzene-1,3-dicarbonate, methyl 5-hydroxy-3-methoxycarbonyl benzoate
Formula Weight 210.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 5-Hydroxyisophthalate