missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-hydroxyisophthalate, 98%

Catalog No. AAB2156530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2156530 250 g
AAB2156509 10 g
AAB2156518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2156530 Supplier Thermo Scientific Chemicals Supplier No. B2156530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13036-02-7 | C10H10O5 | 210.19 g/mol

Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 13036-02-7
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.19
MDL Number MFCD00134367
InChI Key DOSDTCPDBPRFHQ-UHFFFAOYSA-N
PubChem CID 83065
IUPAC Name dimethyl 5-hydroxybenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(O)=C1)C(=O)OC

Specifications

Melting Point 162°C to 164°C
Quantity 250 g
Beilstein 2650120
Synonym dimethyl 5-hydroxyisophthalate, 5-hydroxyisophthalic acid dimethyl ester, 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester, dimethyl5-hydroxyisophthalate, timtec-bb sbb000491, 5-hydroxy-dimethylisophthalate, 1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester, dimethyl-5-hydroxy-benzene-1,3-dicarbonate, methyl 5-hydroxy-3-methoxycarbonyl benzoate
Solubility Information Soluble in water.
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material Dimethyl 5-hydroxyisophthalate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-899-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only