missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate 98.0+%, TCI America™

Catalog No. p-7128700
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52391G 1 g
D52395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52391G Supplier TCI America Supplier No. D52391G

Chemical Identifiers

CAS 944392-68-1
Molecular Formula C16H21BO6
Molecular Weight (g/mol) 320.148
MDL Number MFCD11858596
InChI Key IGSNWXAGFXHYOG-UHFFFAOYSA-N
PubChem CID 42614529
IUPAC Name dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)C(=O)OC)C(=O)OC

Specifications

Melting Point 137°C
Color White
Quantity 1 g
Synonym dimethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalate, 3,5-bis methoxycarbonyl phenylboronic acid pinacol ester, 3,5-bis methoxycarbonyl benzeneboronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-benzenedicarboxylic acid,1,3-dimethyl ester, 1,3-dimethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,3-dicarboxylate, 3,5-dimethyl benzenecarboxylate-5-boronic acid pinacol ester, dimethyl 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl isophthalate, dimethyl5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalate, 1,3-dimethyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,3-dicarboxylate, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalic acid dimethyl ester
Formula Weight 320.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate