missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-Nitrophthalate 98.0+%, TCI America™

Catalog No. D44241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44241G 1 g
D44245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44241G Supplier TCI America Supplier No. D44241G

Chemical Identifiers

CAS 610-22-0
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.18
MDL Number MFCD00017191
InChI Key XWBDWELWBUWSNI-UHFFFAOYSA-N
Synonym dimethyl 4-nitrophthalate, dimethyl4-nitrophthalate, 4-nitrophthalic acid dimethyl ester, dimethyl-4-nitrophthalate, 1,2-benzenedicarboxylic acid, 4-nitro-, dimethyl ester, 1,2-dimethyl 4-nitrophthalate, phthalic acid, 4-nitro-, dimethyl ester, 1,2-benzenedicarboxylic acid, 4-nitro-, 1,2-dimethyl ester, 4-nitrodimethylphthalate, 5-nitroisophthyl acid dimethyl ester
PubChem CID 69119
IUPAC Name 1,2-dimethyl 4-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1C(=O)OC)[N+]([O-])=O

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 195°C
Quantity 1 g
Formula Weight 239.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4-Nitrophthalate