missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 4-Hydroxyisophthalate 97.0+%, TCI America™
$128.62 - $389.59
Chemical Identifiers
| CAS | 5985-24-0 |
|---|---|
| Molecular Formula | C10H10O5 |
| Molecular Weight (g/mol) | 210.185 |
| MDL Number | MFCD00152444 |
| InChI Key | ALBUJVBOIXVVLS-UHFFFAOYSA-N |
| Synonym | 4-Hydroxyisophthalic Acid Dimethyl Ester |
| PubChem CID | 80087 |
| IUPAC Name | dimethyl 4-hydroxybenzene-1,3-dicarboxylate |
| SMILES | COC(=O)C1=CC(=C(C=C1)O)C(=O)OC |
Chemical Identifiers
| 5985-24-0 | |
| 210.185 | |
| ALBUJVBOIXVVLS-UHFFFAOYSA-N | |
| 80087 | |
| COC(=O)C1=CC(=C(C=C1)O)C(=O)OC |
| C10H10O5 | |
| MFCD00152444 | |
| 4-Hydroxyisophthalic Acid Dimethyl Ester | |
| dimethyl 4-hydroxybenzene-1,3-dicarboxylate |
Specifications
| 5985-24-0 | |
| White-Yellow | |
| MFCD00152444 | |
| 4-Hydroxyisophthalic Acid Dimethyl Ester | |
| COC(=O)C1=CC(=C(C=C1)O)C(=O)OC | |
| 210.185 | |
| 210.19 | |
| Crystalline Powder |
| 97°C | |
| C10H10O5 | |
| 1 g | |
| ALBUJVBOIXVVLS-UHFFFAOYSA-N | |
| dimethyl 4-hydroxybenzene-1,3-dicarboxylate | |
| 80087 | |
| ≥97.0% (GC,T) | |
| Dimethyl 4-Hydroxyisophthalate |
Safety and Handling
TSCA : Yes